Learn More
Chemical Identifiers
CAS | 56-84-8 |
---|---|
Molecular Formula | C4H7NO4 |
Molecular Weight (g/mol) | 133.10 |
MDL Number | MFCD00002616 |
InChI Key | CKLJMWTZIZZHCS-UHFFFAOYNA-N |
Synonym | l-aspartic acid, aspartic acid, h-asp-oh, l-aspartate, asparagic acid, aspatofort, l-asparagic acid, l-aminosuccinic acid, s-2-aminosuccinic acid, asparaginic acidShow More |
PubChem CID | 5960 |
ChEBI | CHEBI:17053 |
IUPAC Name | (2S)-2-aminobutanedioic acid |
SMILES | NC(CC(O)=O)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10714981
|
Thermo Scientific Acros
105041000 |
100 g | Plastic Bottle |
£25.07
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10467090
|
Thermo Scientific Acros
105045000 |
500 g | Plastic Bottle |
£47.74
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10341614
|
Thermo Scientific Acros
105040025 |
2.5 kg | Plastic Bottle |
£154.77
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15169153
|
Thermo Scientific Acros
105040100 |
10 kg | Plastic Drum |
£330.65
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
56-84-8 | |
133.10 | |
CKLJMWTZIZZHCS-UHFFFAOYNA-N | |
5960 | |
(2S)-2-aminobutanedioic acid |
C4H7NO4 | |
MFCD00002616 | |
l-aspartic acid, aspartic acid, h-asp-oh, l-aspartate, asparagic acid, aspatofort, l-asparagic acid, l-aminosuccinic acid, s-2-aminosuccinic acid, asparaginic acidShow More | |
CHEBI:17053 | |
NC(CC(O)=O)C(O)=O |
Specifications
>230.0°C | |
56-84-8 | |
200ppm max. | |
Authentic | |
98+% | |
0.5% max. (TLC) | |
HO2CCH2CH(NH2)CO2H | |
+ 25.00 | |
0.1% max. | |
Solubility in water: 5g/L water (25°C). Other solubilities: 2.1g/L water (0°C), 8.5g/L water (40°C), 28.7g/L water (75°C), 16 mg/100 mL ethanol (25°C), soluble in acids, alkalies | |
NC(CC(O)=O)C(O)=O | |
133.10 | |
CHEBI:17053 | |
≥98% | |
L(+)-Aspartic acid, 98+% |
200ppm max. | |
White | |
10ppm max. | |
10ppm max. | |
0.5% max. (105°C, 3 hrs) | |
C4H7NO4 | |
MFCD00002616 | |
300ppm max. | |
l-aspartic acid, aspartic acid, h-asp-oh, l-aspartate, asparagic acid, aspatofort, l-asparagic acid, l-aminosuccinic acid, s-2-aminosuccinic acid, asparaginic acid | |
CKLJMWTZIZZHCS-UHFFFAOYNA-N | |
(2S)-2-aminobutanedioic acid | |
5960 | |
133.1 | |
Crystalline Powder or Crystals |
Safety and Handling
missing translation for 'einecsNumber' : 200-291-6
missing translation for 'rtecsNumber' : CI9098500
missing translation for 'tsca' : TSCA
- L(+)-Aspartic acid
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.