missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Methionine, 98+%

Catalog Number p-156135
22.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
63-68-3
C5H11NO2S
149.21
MFCD00063097 MFCD00801344
FFEARJCKVFRZRR-BYPYZUCNSA-N
l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin
6137
CHEBI:16643
CSCC[C@H](N)C(O)=O
63-68-3
C5H11NO2S
149.21
MFCD00063097 MFCD00801344
FFEARJCKVFRZRR-BYPYZUCNSA-N
l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin
6137
CHEBI:16643
CSCC[C@H](N)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 276.0°C to 279.0°C
Ammonium 300ppm max.
pH 5.5 to 6.5 (aq. soln. 1 in 100)
Color White
Chloride 500ppm max.
Heavy Metals (as Pb) 15ppm max.
Assay Percent Range 98+%
Individual Impurity 0.5% max. of any individual impurity
Infrared Spectrum Authentic
Linear Formula CH3SCH2CH2CH(NH2)CO2H
Iron (Fe) 30ppm max.
Beilstein 04, II, 938
Loss on Drying 0.5% max. (1 g, 105°C)
Merck Index 15, 6047
Packaging Plastic Bottle
Solubility Information Solubility in water: soluble in water: 53.7g/L (20°C). Other solubilities: insoluble in abs.alcohol, ether, petr.ether,, benzene,acetone, ethanol, diethylether, soluble in mineral acids, soluble in warm, dilute alcohol
IUPAC Name (2S)-2-amino-4-(methylsulfanyl)butanoic acid
Specific Rotation + 23.25
Sulfate 300ppm max.
Formula Weight 149.21
Percent Purity ≥98%
Sulfated Ash 0.1% max.
Specific Rotation Condition +23.25° (20°C c=2,6N HCl)
Quantity 25 g
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L-Methionine, 98+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'