missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Phenylalanine, 99% Product Code.: 11488840

Thermo Scientific Chemicals L-Phenylalanine, 99%

Product Code. 11488840
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
1000 g
This item is not returnable. View return policy

Product Code. 11488840

Brand: Thermo Scientific Chemicals A13238.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L-Phenylalanine, CAS # 63-91-2, is an essential amino acid with an aromatic structure used in the biosynthesis of other amino acids. It is important for the structure and function of many proteins and enzymes.

L-Phenylalanine is an aromatic organic compound containing amine and acid as functional groups. It is an essential amino acid and its structure can be considered as the derivative of 3-phenylpropionic acid, in which one hydrogen atom present at C-2 position is substituted with amino group. It is a chiral compound and consists of two optical isomers. It is also known as 2-amino-3-phenylpropanoic acid, beta-phenylalanine, L-alfa-amino-beta-phenylpropionic acid, H-Phe-OH and (S)-2-amino-3-phenylpropionic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • L-Phenylalanine is an essential amino acid found in several dietary sources, including meat, fish, eggs, cheese, and milk
  • Phenylalanine can be converted into the amino acid tyrosine

Application

  • L-Phenylalanine is used in the synthesis of other amino acids
  • It can be used as a precursor for the skin pigment melanin as well as the monoamine neurotransmitters dopamine, norepinephrine (noradrenaline), and epinephrine (adrenaline)
  • It is used to biochemically form proteins coded by DNA
  • Due to its ability to absorb ultraviolet radiation, it is used to quantify the amount of protein
  • In research, it is used as an alkaline phosphatase inhibitor
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-91-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00064227
InChI Key COLNVLDHVKWLRT-UHFFFAOYNA-N
Synonym l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-ohShow MoreShow Less
PubChem CID 6140
ChEBI CHEBI:17295
SMILES NC(CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point ∼275°C (decomposition)
Assay Percent Range 99%
Beilstein 1910408
Merck Index 14,7271
Solubility Information Soluble in water. Slightly soluble in methanol and ethanol. Insoluble in ethyl ether and benzene.
IUPAC Name (2S)-2-amino-3-phenylpropanoic acid
Quantity 500 g
Formula Weight 165.19
Optical Rotation −34° (c=2 in Water)
Percent Purity 99%
Chemical Name or Material L-Phenylalanine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252961 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog NumberA13238.18,A13238.22,A13238.0B,A13238.14,A13238.36
Lot Number10249299 Certificate TypeCertificate of Analysis Date02/02/2024 Catalog NumberA13238.18,A13238.22,A13238.0B,A13238.14,A13238.36
Lot Number10246845 Certificate TypeCertificate of Analysis Date19/10/2023 Catalog NumberA13238.18,A13238.22,A13238.0B,A13238.14,A13238.36
Lot Number10221473 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13238.18,A13238.22,A13238.0B,A13238.14,A13238.36
Lot NumberP07C028 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13238.18,A13238.22,A13238.0B,A13238.14,A13238.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Phenylalanine, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.