missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Serine, 99% Product Code.: 11409123

Thermo Scientific Chemicals L-Serine, 99%

Product Code. 11409123
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11409123

Brand: Thermo Scientific Chemicals A11179.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L-serine is used as a dietary supplement to improve health under certain conditions of malnutrition or physical stress, as a cosmetic incredient, as ingredients for cell culture media. It is used as a precursor to several amino acids, including glycine, cysteine and other metabolites such as sphingolipids and folate. It is used in bio-synthesis of purines and pyrimidines. It plays a vital role in catalytic function of enzymes. Further, L-serine and its metabolic products are essential for cell proliferation and specific functions of the central nervous system. .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-serine is used as a dietary supplement to improve health under certain conditions of malnutrition or physical stress, as a cosmetic incredient, as ingredients for cell culture media. It is used as a precursor to several amino acids, including glycine, cysteine and other metabolites such as sphingolipids and folate. It is used in bio-synthesis of purines and pyrimidines. It plays a vital role in catalytic function of enzymes. Further, L-serine and its metabolic products are essential for cell proliferation and specific functions of the central nervous system. .

Solubility
Soluble in water. Insoluble in ethanol, ether and benzene.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-45-1
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.093
MDL Number MFCD00064224
InChI Key MTCFGRXMJLQNBG-REOHCLBHSA-N
Synonym l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acidShow MoreShow Less
PubChem CID 5951
ChEBI CHEBI:17115
IUPAC Name (2S)-2-amino-3-hydroxypropanoic acid
SMILES C(C(C(=O)O)N)O

Specifications

Melting Point 214°C to 224°C (decomposition)
Beilstein 1721404
Merck Index 14,8460
Solubility Information Soluble in water. Insoluble in ethanol,ether and benzene.
Quantity 500 g
Formula Weight 105.09
Optical Rotation +15° (c=9 in 1N HCl)
Percent Purity 99%
Chemical Name or Material L-Serine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10248001 Certificate TypeCertificate of Analysis Date27/09/2023 Catalog NumberA11179.14,A11179.36,A11179.22
Lot Number10244269 Certificate TypeCertificate of Analysis Date01/11/2022 Catalog NumberA11179.14,A11179.36,A11179.22
Lot Number10145845 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11179.14,A11179.36,A11179.22
Lot Number5025A12W Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11179.14,A11179.36,A11179.22
Lot Number10158016 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11179.14,A11179.36,A11179.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Serine, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.