missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-(-)-threo-3-Hydroxyaspartic acid, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Description
L-(-)-threo-3-Hydroxyaspartic acid is a potent, competitive, transportable EAAT1-4 inhibitor/non-transportable EAAT5 inhibitor.
Specifications
Specifications
CAS | 7298-99-9 |
Molecular Formula | C4H7NO5 |
Synonym | dl-threo-beta-hydroxyaspartic acid, unii-z9q5w0u8vw, 2s,3s-2-amino-3-hydroxybutanedioic acid, l---threo-3-hydroxyaspartic acid, z9q5w0u8vw, dl-threo-beta-hydroxyaspartate, 3s-3-hydroxy-l-aspartic acid, d,l-threo-beta-hydroxy aspartic acid, 3-hydroxyaspartic acid, beta-hydroxyaspartic acid |
InChI Key | YYLQUHNPNCGKJQ-LWMBPPNESA-N |
SMILES | C(C(C(=O)O)O)(C(=O)O)N |
IUPAC Name | (2S,3S)-2-amino-3-hydroxybutanedioic acid |
Molecular Weight (g/mol) | 149.102 |
Quantity | 10 mg |
PubChem CID | 443239 |
ChEBI | CHEBI:10696 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
L-(-)-threo-3-Hydroxyaspartic acid, Tocris Bioscience™ >
Spot an opportunity for improvement?Share a Content Correction