missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Phosphate Buffered Saline (25X)
Dilute Epredia™ Lab Vision™ Phosphate Buffered Saline (25X) before use to make stock solution, which acts as an ideal wash buffer for immunolabeling techniques.
Brand: Epredia™ TA-125-PB
This item is not returnable.
View return policy
Description
- For in vitro diagnostic use
Storage Conditions: Store at room temperature. Stable for up to two years. Use diluted solution within one week.

Specifications
125 mL | |
25 X | |
125 mL | |
Healthcare |
Room temperature | |
Phosphate Buffered Saline (PBS) | |
Phosphate Buffered Saline |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
74532514 | Certificate of Analysis | 29/01/2025 | TA-125-PB |
74689831 | Certificate of Analysis | 29/01/2025 | TA-125-PB |
74710053 | Certificate of Analysis | 29/01/2025 | TA-125-PB |
74606449 | Certificate of Analysis | 29/01/2025 | TA-125-PB |
74710053 | Certificate of Analysis | 29/07/2022 | TA-125-PB |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Epredia™ Lab Vision™ Phosphate Buffered Saline (25X) > 125mL
Spot an opportunity for improvement?Share a Content Correction