missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lab Vision™ Tris-Buffered Saline (25X), Epredia™
Take advantage of a wash buffer for immunolabeling techniques with the Epredia™ Lab Vision™ Tris-Buffered Saline (25X),
Brand: Epredia TA-125-TB
This item is not returnable.
View return policy
Description
Storage Conditions: Store at room temperature. Stable for 2 years from date of manufacture. Use diluted solution within one week.
Specifications
Tris-Buffered Saline (25X) | |
25X | |
Healthcare | |
125 mL |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lab Vision™ Tris-Buffered Saline (25X), Epredia™ > 125mL
For in vitro diagnostic use
Spot an opportunity for improvement?Share a Content Correction