missing translation for 'onlineSavingsMsg'
Learn More

Laboratory Buffer Solution, pH 2.00, Reagecon™
BIOPHARMA

Designed for use in laboratory pH measurement. Reagecon™ Laboratory Buffer Solution, pH 2.00 is tested at 20°C to an accuracy level of ±0.02. This calibration and control buffer is specially formulated to ensure its stability and is supplied with a detailed Certificate of Analysis.

Brand:  Reagecon 90518

Product Code. 11726398

  • £34.90 / 250mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Reagecon manufactures the most comprehensive range of pH reagents in the world; these are designed to suit all end user requirements. This laboratory grade buffer is manufactured to exacting specifications with an extended shelf life and is supplied with a detailed Certificate of Analysis and comprehensive label.
Specifications

Specifications

0°C
2.00 ±0.02
250 mL
Fully miscible in water
100°C
Liquid
Characteristic
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2C24G1 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog Number90518
Lot Number2C23M1 Certificate TypeCertificate of Analysis Date14/01/2025 Catalog Number90518
Lot Number2C22H1 Certificate TypeCertificate of Analysis Date16/03/2024 Catalog Number90518
Lot Number2C20M1 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number90518
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Laboratory Buffer Solution, pH 2.00, Reagecon™ > Quantity: 250mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.