Learn More
Bohlender™ SICCO™ Auto Glove Box with Transfer Chamber
Auto Glove Box with Transfer Chamber made of acrylic glass, aluminum, and polyethylene.
Brand:  Bohlender™ V1755-08
Product Code. 17137684
  • £12320.00 / Each
Explore more special offers
Description
Description
  • Aluminum frame with panels made of acrylic glass, base panel made of polyethylene
  • Front panel with two glove ports and one pair of natural rubber gloves size 9
  • Two lead-ins for cables on the back side (upper left and upper right), including one hygrometer in the working space and the transferchamber each
  • Transfer chamber with two doors, external door with latch to prevent accidental opening
  • Tray extends into the workspace with opening of the internal door for easy removal of inserted product
  • Automatic reduction of humidity inside the workspace and transfer chamber onto below 20% through automatic drying units mounted on the back side of the workspace and desiccant cartridges in the outer wall of the transfer chamber
  • Desiccant cartridges can be exchanged for regeneration in case of saturation
  • Electrical power connection 230 V / 50 Hz is required
  • Robust and light-weight aluminum construction
  • Superb all-round visibility
  • Non-slip rubber feet
  • Cable lead-ins for electric devices
  • Large front opening
  • Automatic drying (manual regeneration of desiccant is not necessary)
  • Constant low interior humidity between 20% and 30%
  • Transfer chamber for inserting further products
Specifications
Specifications
Acrylic Glass, Aluminum, Polyethylene
21 in.
540 mm
26 in.
650 mm
44 lb.
48 in.
1210 mm
24 in.
600 mm
28 in.
700 mm
290 L
20 kg
33 in.
850 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Bohlender™ SICCO™ Auto Glove Box with Transfer Chamber > Auto Glove Box with Transfer Chamber; Material: Acrylic Glass, Aluminum, Polyethylene; Capacity: 290 L; Depth (Metric) Exterior: 600 mmHeight (Metric) Exterior: 700 mm; Width (Metric) Exterior: 1210 mm