Learn More
Thermo Scientific™ Labtip Pipette Tips
GREENER_CHOICE
Basic pipette tips for standard liquid transfer needs.
£43.30 - £442.95
Specifications
Filtered Non-filtered
Sterility Non-sterile
Product Line LabTip
Material Virgin Polypropylene
View More Specs
Products 4
Product Code Brand Volume (Metric) Format Packaging Price Quantity & Availability  
Product Code Brand Volume (Metric) Format Packaging Price Quantity & Availability  
12326582
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
94300210
1000 μL Bulk 10,000 tips/bag
£213.00
Pack of 10000
In Stock
to see stock.
Add to basket
12306582
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
94300110
200 μL Bulk 25,000 tips/bag
£442.95
Pack of 25000
Estimated Shipment: 28-05-2025
to see stock.
Add to basket
10122602
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
94300120
200 μL Bulk 1000 tips/bag
£43.30
Pack of 1000
In Stock
to see stock.
Edge
Add to basket
12316582
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
94300130
200 μL Rack 10 x 96/Tray
£268.42
Pack of 3840
In Stock
to see stock.
Add to basket
Description
Description
Thermo Scientific™ Labtip standard pipette tips are made from 100% virgin polypropylene. These tips are available in 10, 200, 300 and 1000 μL sizes. Available in clear (10 μL and 300 μL), yellow (200 μL), blue (1000 μL), these tips for general lab use across countless liquid transfer applications.
Labtip pipette tips provide:
  • The most common sizes used in a lab: 10, 200, 300 and 1000 μL
  • Non-Filter essential tips for general laboratory usage
  • Non-sterile packaging in both Racked and Bulk options
Packaging Options:
  • Racked, nonsterile - 96 tips/ tray, 10 trays/pack
  • Bulk - 1,000 tips/ bag
  • Bulk - 25,000 tips/bag
Specifications
Specifications
Non-filtered
Non-sterile
LabTip
Virgin Polypropylene
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Labtip Pipette Tips