missing translation for 'onlineSavingsMsg'
Learn More
Lanthanum(III) bromide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11468590

Lanthanum(III) bromide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11468590
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11468590

Brand: Thermo Scientific Alfa Aesar 044396.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13536-79-3
Molecular Formula Br3La
Molecular Weight (g/mol) 378.62
MDL Number MFCD00016161
InChI Key XKUYOJZZLGFZTC-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) tribromide
SMILES [Br-].[Br-].[Br-].[La+3]

Specifications

Packaging Ampouled under argon
Quantity 10 g
Formula Weight 378.62
Percent Purity 99.9%
Physical Form Lumps
Chemical Name or Material Lanthanum(III) bromide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU06K010 Certificate TypeCertificate of Analysis Date22/08/2024 Catalog Number044396.18,044396.09
Lot NumberU06K012 Certificate TypeCertificate of Analysis Date22/08/2024 Catalog Number044396.18,044396.09
Lot NumberP07K002 Certificate TypeCertificate of Analysis Date13/03/2024 Catalog Number044396.18,044396.09
Lot NumberP07K003 Certificate TypeCertificate of Analysis Date13/03/2024 Catalog Number044396.18,044396.09
Lot NumberP01I024 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number044396.18,044396.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lanthanum(III) bromide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.