Learn More
Lanthanum(III) carbonate hydrate, 99.9% (REO), Thermo Scientific Chemicals
Description
It is used in pharmaceuticals & used in medicine as a phosphate binder.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Especificaciones
Physical Form | Powder |
Quantity | 25 g |
Merck Index | 14,5364 |
Solubility Information | Insoluble in water. |
Formula Weight | 457.85 (Anhydrous) |
Percent Purity | 99.9% |
Sensitivity | Hygroscopic |
Density | 2.6 g/mL to 2.7 g/mL |
Assay | (REO) |
Chemical Name or Material | Lanthanum(III) carbonate hydrate |
Sugerencias de productos
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
T11K014 | Certificate of Analysis | 26/07/2024 | 016639.14,016639.36,016639.22 |
Q18K027 | Certificate of Analysis | 23/04/2024 | 016639.14,016639.36,016639.22 |
R01F039 | Certificate of Analysis | 07/12/2021 | 016639.14,016639.36,016639.22 |
D13P28 | Certificate of Analysis | 07/12/2021 | 016639.14,016639.36,016639.22 |
C24T051 | Certificate of Analysis | 07/12/2021 | 016639.14,016639.36,016639.22 |
RUO – Research Use Only
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.