missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) carbonate hydrate, 99.9% (REO), Thermo Scientific Chemicals

£18.69 - £155.78

Chemical Identifiers

CAS 54451-24-0
Molecular Formula C3La2O9
Molecular Weight (g/mol) 457.84
MDL Number MFCD00011069
InChI Key NZPIUJUFIFZSPW-UHFFFAOYSA-H
IUPAC Name dilanthanum(3+) tricarbonate
SMILES [La+3].[La+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11357198
View Documents
Thermo Scientific Alfa Aesar
016639.14
25 g
£18.69
25g
Please to purchase this item. Need a web account? Register with us today!
11347198
View Documents
Thermo Scientific Alfa Aesar
016639.22
100 g
£45.23
100g
Please to purchase this item. Need a web account? Register with us today!
11337198
View Documents
Thermo Scientific Alfa Aesar
016639.36
500 g
£155.78
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used in pharmaceuticals & used in medicine as a phosphate binder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

54451-24-0
457.84
NZPIUJUFIFZSPW-UHFFFAOYSA-H
[La+3].[La+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
C3La2O9
MFCD00011069
dilanthanum(3+) tricarbonate

Specifications

54451-24-0
C3La2O9
14,5364
NZPIUJUFIFZSPW-UHFFFAOYSA-H
dilanthanum(3+) tricarbonate
457.85 (Anhydrous)
Hygroscopic
(REO)
Powder
MFCD00011069
Insoluble in water.
[La+3].[La+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
457.84
99.9%
2.6 g/mL to 2.7 g/mL
Lanthanum(III) carbonate hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 209-599-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only