missing translation for 'onlineSavingsMsg'
Learn More
Lanthanum(III) chloride hydrate, Honeywell™ Product Code.: 15626040

Lanthanum(III) chloride hydrate, Honeywell™

Product Code. 15626040 Shop All Honeywell Research Chemicals Products
500 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 15626040

Brand: Honeywell Research Chemicals 61490500G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

∼7 mol/mol water,≥98.5% (calc. based on dry substance, AT)

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20211-76-1
Molecular Formula Cl3La
Molecular Weight (g/mol) 245.26
MDL Number MFCD00149753
InChI Key ICAKDTKJOYSXGC-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[La+3]

Specifications

Quantity 500 g
Linear Formula LaCl3 · xH2O
UN Number NONH for all modes of transport
Formula Weight 245.26g/mol (anhydrous basis)
Percent Purity ≥98.5% (calculated based on dry substance, AT)
Chemical Name or Material Lanthanum(III) chloride hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lanthanum(III) chloride hydrate, Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.