missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Lawesson's Reagent, 97% Product Code.: 11401898

Thermo Scientific Chemicals Lawesson's Reagent, 97%

Product Code. 11401898
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11401898

Brand: Thermo Scientific Chemicals A14530.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lawesson's reagent is a thiation agent used to convert carbonyl compounds into thiocarbonyls. It is also used to thionate enones, esters, lactones, amides, lactams and quinones. Further, it is used to prepare thiols from alcohols. It is associated with silver perchlorate and utilized as and oxophilic Lewis acid catalyst for Diels-Alder reaction of dienes with alfa, beta-unsaturated aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lawesson′s reagent is a thiation agent used to convert carbonyl compounds into thiocarbonyls. It is also used to thionate enones, esters, lactones, amides, lactams and quinones. Further, it is used to prepare thiols from alcohols. It is associated with silver perchlorate and utilized as and oxophilic Lewis acid catalyst for Diels-Alder reaction of dienes with alfa, beta-unsaturated aldehydes.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19172-47-5
Molecular Formula C14H14O2P2S4
Molecular Weight (g/mol) 404.452
MDL Number MFCD00005171
InChI Key CFHGBZLNZZVTAY-UHFFFAOYSA-N
Synonym lawesson's reagent, lawesson reagent, 2,4-bis 4-methoxyphenyl-1,3,2,4-dithiadiphosphetane 2,4-disulfide, unii-a4125mq8rx, 1,3,2,4-dithiadiphosphetane, 2,4-bis 4-methoxyphenyl-, 2,4-disulfide, 2,4-bis 4-methoxyphenyl-2,4-dithioxo-1,3,2,4-dithiadiphosphetane, 2,4-bis 4-methoxyphenyl-1,3,2,4-dithiadiphosphetane-2,4-disulfide, 2,4-bis 4-methoxyphenyl-1,3-dithia-2,4-diphosphetane 2,4-disulfide, 2,4-bis 4-methoxyphenyl-1,3-dithia-2,4-diphosphetane 2,4-disulphide, 4-methoxyphenylthiophosphoric cyclic di thioanhydrideShow MoreShow Less
PubChem CID 87949
IUPAC Name 2,4-bis(4-methoxyphenyl)-2,4-bis(sulfanylidene)-1,3,2$l^{5},4$l^{5}-dithiadiphosphetane
SMILES COC1=CC=C(C=C1)P2(=S)SP(=S)(S2)C3=CC=C(C=C3)OC

Specifications

Melting Point 221°C to 225°C
Quantity 25 g
UN Number UN3134
Beilstein 1024888
Sensitivity Moisture sensitive
Merck Index 14,5391
Solubility Information Insoluble in water.
Formula Weight 404.47
Percent Purity 97%
Chemical Name or Material Lawesson's Reagent

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10238993 Certificate TypeCertificate of Analysis Date07/12/2023 Catalog NumberA14530.14,A14530.36,A14530.22,A14530.06
Lot Number10233049 Certificate TypeCertificate of Analysis Date11/07/2023 Catalog NumberA14530.14,A14530.36,A14530.22,A14530.06,A14530
Lot Number10179911 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14530.14,A14530.36,A14530.22,A14530.06
Lot Number10220993 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14530.14,A14530.36,A14530.22,A14530.06
Lot Number10177164 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14530.14,A14530.36,A14530.22,A14530.06
compliance-icons
Product Identifier
  • Lawesson's Reagent
Signal Word
  • Danger
Hazard Category
  • Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
  • H261-In contact with water releases flammable gases.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : 1,3,2,4-Dithiadiphosphetane, 2,4-bis(4-methoxyphenyl)-, 2,4-disulfide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Lawesson's Reagent, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.