missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(II) carbonate, basic, extra pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
500 g
2.5 kg
10 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 400.0°C |
Color | White |
Physical Form | Powder |
Quantity | 2.5 kg |
Linear Formula | (PbCO3)2·Pb(OH)2 |
Merck Index | 15, 1002 |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in acids |
Formula Weight | 775.59 |
Percent Purity | ≥78% (Pb) |
Grade | Extra Pure |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0442443 | Certificate of Analysis | 31/01/2025 | 222715000,222710025 |
A0459848 | Certificate of Analysis | 15/02/2024 | 222715000,222710050,222710025,222710100 |
A0455926 | Certificate of Analysis | 07/09/2023 | 222715000,222710050,222710025,222710100 |
A0422454 | Certificate of Analysis | 28/08/2023 | 222715000,222710050,222710025 |
A0451657 | Certificate of Analysis | 02/05/2023 | 222715000,222710025 |
Safety and Handling



Product Identifier
- Lead(II) carbonate, basic
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lead(II) carbonate, basic, extra pure, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction