missing translation for 'onlineSavingsMsg'
Learn More
Lead(II) cyanurate monohydrate, Thermo Scientific™ Product Code.: 11308067

Lead(II) cyanurate monohydrate, Thermo Scientific™

Product Code. 11308067
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11308067

Brand: Thermo Scientific Alfa Aesar 022556.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53846-29-0
Molecular Formula C6H6N6O9Pb3
Molecular Weight (g/mol) 927.747
MDL Number MFCD00064816
InChI Key DPLKIANDZYQWKF-UHFFFAOYSA-H
Synonym lead cyanurate, lead ii cyanurate monohydrate, lead 2+ ;1,3,5-triazine-2,4,6-triolate;trihydrate
PubChem CID 90473286
IUPAC Name lead(2+);1,3,5-triazine-2,4,6-triolate;trihydrate
SMILES C1(=NC(=NC(=N1)[O-])[O-])[O-].C1(=NC(=NC(=N1)[O-])[O-])[O-].O.O.O.[Pb+2].[Pb+2].[Pb+2]

Specifications

Quantity 5 g
UN Number UN3467
Formula Weight 891.69 (873.67 Anhydrous)
Physical Form Powder
Chemical Name or Material Lead(II) cyanurate monohydrate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) cyanurate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lead(II) cyanurate monohydrate, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.