missing translation for 'onlineSavingsMsg'
Learn More
Lead(II) telluride, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11319239

Lead(II) telluride, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11319239
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11319239

Brand: Thermo Scientific Alfa Aesar 088285.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lead(II) telluride is used to make thermoelectric converter (cooler, generator, heat), infrared detectors and tunnel diode. It is also an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-91-6
Molecular Formula PbTe
Molecular Weight (g/mol) 334.80
MDL Number MFCD00016274
InChI Key OCGWQDWYSQAFTO-UHFFFAOYSA-N
Synonym lead telluride, lead ii telluride, unii-v1og6oa4bj, v1og6oa4bj, plumbanetellone, bleitellurid, lead monotelluride, lead ii telluride trace metals basisShow MoreShow Less
PubChem CID 4389803
IUPAC Name tellanylidenelead
SMILES [Te]=[Pb]

Specifications

Melting Point 917°C
Physical Form Pieces
Quantity 25 g
UN Number UN3284
Merck Index 14,5422
Solubility Information Insoluble in water.
Formula Weight 334.79
Percent Purity 99.999%
Odor Product specific
Density 8.164 g/mL
Assay (metals basis)
Chemical Name or Material Lead (II) telluride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ15K045 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number088285.14,088285.22
Lot NumberA24N06 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number088285.14,088285.22
Lot NumberL08P07 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number088285.14,088285.22
Lot NumberG15U027 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number088285.14,088285.22
Lot NumberE06Z042 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number088285.14,088285.22
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) telluride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lead(II) telluride, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.