Learn More
Lead(II) titanium oxide, 99.5%, Thermo Scientific Chemicals
£38.40 - £183.00
Chemical Identifiers
CAS | 12060-00-3 |
---|---|
Molecular Formula | O3PbTi |
Molecular Weight (g/mol) | 303.064 |
MDL Number | MFCD00011159 |
InChI Key | NKZSPGSOXYXWQA-UHFFFAOYSA-N |
Synonym | lead titanate, lead titanium trioxide, lead ii titanate, lead titanium oxide, lead titanium oxide pbtio3, titanic acid, lead salt, titanox l, titanicacid,leadsalt, o3ti.pb, lead titanate, powder, |
PubChem CID | 16211560 |
IUPAC Name | dioxido(oxo)titanium;lead(2+) |
SMILES | [O-][Ti](=O)[O-].[Pb+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11387737
|
Thermo Scientific Alfa Aesar
014139.22 |
100 g |
£38.40
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11397737
|
Thermo Scientific Alfa Aesar
014139.36 |
500 g |
£183.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11307747
|
Thermo Scientific Alfa Aesar
014139.A3 |
2 kg |
£160.00
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lead(II) titanium oxide is widely used in ceramic capacitor, ceramic filters, PTC thermal resistor, pressure sensitive resistor and the performance of transducer field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12060-00-3 | |
303.064 | |
NKZSPGSOXYXWQA-UHFFFAOYSA-N | |
16211560 | |
[O-][Ti](=O)[O-].[Pb+2] |
O3PbTi | |
MFCD00011159 | |
lead titanate, lead titanium trioxide, lead ii titanate, lead titanium oxide, lead titanium oxide pbtio3, titanic acid, lead salt, titanox l, titanicacid,leadsalt, o3ti.pb, lead titanate, powder, | |
dioxido(oxo)titanium;lead(2+) |
Specifications
12060-00-3 | |
Powder | |
MFCD00011159 | |
lead titanate, lead titanium trioxide, lead ii titanate, lead titanium oxide, lead titanium oxide pbtio3, titanic acid, lead salt, titanox l, titanicacid,leadsalt, o3ti.pb, lead titanate, powder, | |
NKZSPGSOXYXWQA-UHFFFAOYSA-N | |
dioxido(oxo)titanium;lead(2+) | |
16211560 | |
99.5% | |
7.520 g/mL |
Yellow | |
O3PbTi | |
UN3288 | |
Insoluble in water. | |
[O-][Ti](=O)[O-].[Pb+2] | |
303.064 | |
303.09 | |
Odorless | |
Lead(II) titanium oxide |
Safety and Handling
GHS H Statement
H300-H360-H373-H332
Fatal if swallowed.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
P201-P202-P260-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P330-P501c
H300-H332-H350-H360Df-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 235-038-9
missing translation for 'rtecsNumber' : XR1575000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Lead(II) titanium oxide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H300-Fatal if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P201-Obtain special instructions before use.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only