missing translation for 'onlineSavingsMsg'
Learn More
Lead(IV) oxide, 97+%, extra pure, Thermo Scientific Chemicals Product Code.: 10480681

Lead(IV) oxide, 97+%, extra pure, Thermo Scientific Chemicals

Product Code. 10480681
500 g, Glass bottle
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10480681

Brand: Thermo Scientific Acros 217535000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-60-0
Molecular Formula O2Pb
Molecular Weight (g/mol) 239.20
MDL Number MFCD00011165
InChI Key YADSGOSSYOOKMP-UHFFFAOYSA-N
Synonym lead dioxide, lead iv oxide, lead peroxide, lead superoxide, lead oxide pbo2, lead brown, lead oxide brown, lead peroxide pbo2, bioxyde de plomb frenchShow MoreShow Less
PubChem CID 14793
SMILES O=[Pb]=O

Specifications

Melting Point 290°C
Color Brown to Black
Physical Form Powder
Quantity 500 g
Assay Percent Range 97+%
Linear Formula PbO2
Fieser 01,5331; 02,233; 03,168; 11,282
Merck Index 15, 5462
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hcl and glacial acetic acid,insoluble in alcohol
IUPAC Name dioxolead
Formula Weight 239.19
Percent Purity ≥97%
Grade Extra Pure
Packaging Glass bottle
Chemical Name or Material Lead(IV) oxide, p.a.
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0442042 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number217531000,217530050,217535000,217530025
Lot NumberA0414657 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number217531000,217530050,217535000,217530025
Lot NumberA0358837 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number217531000,217530050,217535000,217530025
Lot NumberA0442042 Certificate TypeCertificate of Analysis Date17/05/2022 Catalog Number217531000,217530050,217535000,217530025
Lot NumberA0432145 Certificate TypeCertificate of Analysis Date15/07/2021 Catalog Number217531000,217530050,217535000,217530025

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lead(IV) oxide, 97+%, extra pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.