missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Levamisole hydrochloride, 99+% Product Code.: 10666202

Thermo Scientific Chemicals Levamisole hydrochloride, 99+%

Product Code. 10666202
10 g, Glass bottle
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10666202

Brand: Thermo Scientific Chemicals 187870100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16595-80-5
Molecular Formula C11H12N2S·ClH
Molecular Weight (g/mol) 240.76
MDL Number MFCD00012675
InChI Key LAZPBGZRMVRFKY-HNCPQSOCSA-N
Synonym levamisole hydrochloride, levamisole hcl, ergamisol, tramisol, tramisole, decaris, --tetramisole hydrochloride, niratic hydrochloride, ascaridil, dekarisShow MoreShow Less
PubChem CID 27944
ChEBI CHEBI:6433
IUPAC Name (6S)-6-phenyl-2,3,5,6-tetrahydroimidazo[2,1-b][1,3]thiazole;hydrochloride
SMILES C1CSC2=NC(CN21)C3=CC=CC=C3.Cl

Specifications

Melting Point 226°C to 231°C
Color White
Loss on Drying 0.5% max. (1 g, 105°C, 4 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 10 g
Merck Index 15, 5511
Specific Rotation Condition − 127.50 (20.00°C c=5,H2O)
Specific Rotation -127.5
Solubility Information Solubility in water: 210g/L (20°C). Other solubilities: soluble in methanol and propylene glycol,sparingly soluble in ethanol,slightly soluble in chloroform,hexane and acetone
Formula Weight 240.76
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Levamisole hydrochloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468162 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number187870500,187870100
Lot NumberA0465908 Certificate TypeCertificate of Analysis Date06/09/2024 Catalog Number187870500,187870100
Lot NumberA0444417 Certificate TypeCertificate of Analysis Date27/02/2023 Catalog Number187870500,187870100
Lot NumberA0395278 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number187870500,187870100
Lot NumberA0405532 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number187870500,187870100
compliance-icons
Product Identifier
  • Levamisole hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Levamisole hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Levamisole hydrochloride, 99+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.