missing translation for 'onlineSavingsMsg'
Learn More
Levulinic Acid, 98+%, Thermo Scientific Chemicals Product Code.: 10225080

Levulinic Acid, 98+%, Thermo Scientific Chemicals

Product Code. 10225080
1 kg, Glass bottle
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10225080

Brand: Thermo Scientific Acros 125140010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-76-2
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.12
MDL Number MFCD00002796
InChI Key JOOXCMJARBKPKM-UHFFFAOYSA-N
Synonym levulinic acid, laevulinic acid, pentanoic acid, 4-oxo, levulic acid, 4-oxovaleric acid, 3-acetylpropionic acid, 4-ketovaleric acid, leva, gamma-ketovaleric acid, acetopropionic acidShow MoreShow Less
PubChem CID 11579
ChEBI CHEBI:45630
IUPAC Name 4-oxopentanoic acid
SMILES CC(=O)CCC(=O)O

Specifications

Melting Point 33.0°C to 35.0°C
Color Yellow
Density 1.1300g/mL
Boiling Point 245.0°C to 246.0°C
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula CH3COCH2CH2CO2H
Refractive Index 1.4396 to 1.4426
Quantity 1 kg
Beilstein 03, 671
Fieser 01,564; 02,239; 10,230
Merck Index 15, 5526
Specific Gravity 1.13
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol and ether,insoluble in aliphatic hydrocarbons
Formula Weight 116.12
Percent Purity 98+%
Physical Form Liquid After Melting
Chemical Name or Material Levulinic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0457755 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number125141000,125140010,125142500,125140050
Lot NumberA0462749 Certificate TypeCertificate of Analysis Date16/05/2024 Catalog Number125140010,125141000,125142500,125140050
Lot NumberA0451233 Certificate TypeCertificate of Analysis Date25/03/2024 Catalog Number125140010,125141000,125142500,125140050
Lot NumberA0459760 Certificate TypeCertificate of Analysis Date12/02/2024 Catalog Number125140010,125141000,125142500,125140050
Lot NumberA0457755 Certificate TypeCertificate of Analysis Date17/11/2023 Catalog Number125140010,125141000,125142500,125140050
compliance-icons
Product Identifier
  • Levulinic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Levulinic Acid, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.