missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Purolite™ Lifetech™ ECR Resins for Enzyme Immobilization, 300 to 710 μm
Lifetech™ ECR resins are robust polymeric resins designed for research and industrial immobilization of enzymes. Available for covalent, ionic, adsorption and affinity immobilization.
Brand: Purolite™ LS01395-192
This item is not returnable.
View return policy
Description
- Enzyme immobilization by adsorption, covalent, IEX and affinity
- High mechanical stability
- High porosity
- Easily dried after immobilization
- Suitable for batch and column configuration
- Optimal for use in organic solvent/water free systems
- Compliant with ResAP(2004) 3
- Halal, Kosher and TSE/BSE/GMO free
Specifications
White to off-white spherical beads | |
1200 to 1800 Å | |
300 to 710 μm | |
Amino | |
Lifetech™ | |
Methacrylate (ECR Resin) |
3 to 10 | |
2°C to 20°C | |
70 to 80% | |
Wet | |
250 g |