missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Purolite™ Lifetech™ ECR Resins for Enzyme Immobilization, 300 to 710 μm
Lifetech™ ECR resins are robust polymeric resins designed for research and industrial immobilization of enzymes. Available for covalent, ionic, adsorption and affinity immobilization.
£41.75 - £627.00
Specifications
Description | White to off-white spherical beads |
---|---|
Particle Size | 300 to 710 μm |
Form | Wet |
Product Line | Lifetech™ |
Type | Methacrylate (ECR Resin) |
Product Code | Brand | Product Line | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Line | Quantity | Price | Quantity & Availability | ||||
17009343
|
Purolite™
LS01245-94 |
Lifetech™ | 1 kg | N/A | |||||
17009373
|
Purolite™
LS01375-94 |
Lifetech™ | 1 kg | N/A | |||||
17019323
|
Purolite™
LS01125-192 |
Lifetech™ | 250 g | N/A | |||||
17019383
|
Purolite™
LS01395-192 |
Lifetech™ | 250 g | N/A | |||||
17029323
|
Purolite™
LS01125-94 |
Lifetech™ | 1 kg | N/A | |||||
17029383
|
Purolite™
LS01395-94 |
Lifetech™ | 1 kg | N/A | |||||
17039333
|
Purolite™
LS01235-192 |
Lifetech™ | 250 g | N/A | |||||
17039363
|
Purolite™
LS01365-192 |
Lifetech™ | 250 g | N/A | |||||
17049333
|
Purolite™
LS01235-94 |
Lifetech™ | 1 kg | N/A | |||||
17049353
|
Purolite™
LS01355-192 |
Lifetech™ | 250 g | N/A | |||||
17049363
|
Purolite™
LS01365-94 |
Lifetech™ | 1 kg | N/A | |||||
17049383
|
Purolite™
LS01415-192 |
Lifetech™ | 250 g | N/A | |||||
17059353
|
Purolite™
LS01355-94 |
Lifetech™ | 1 kg | N/A | |||||
17059373
|
Purolite™
LS01385-192 |
Lifetech™ | 250 g | N/A | |||||
17059383
|
Purolite™
LS01415-94 |
Lifetech™ | 1 kg | N/A | |||||
17069373
|
Purolite™
LS01385-94 |
Lifetech™ | 1 kg | N/A | |||||
17079313
|
Purolite™
LS01105-186 |
Lifetech™ | 50 g | N/A | |||||
17079323
|
Purolite™
LS01215-192 |
Lifetech™ | 250 g | N/A | |||||
17079353
|
Purolite™
LS01356-192 |
Lifetech™ | 250 g | N/A | |||||
17089313
|
Purolite™
LS01105-192 |
Lifetech™ | 250 g | N/A | |||||
17089323
|
Purolite™
LS01215-94 |
Lifetech™ | 1 kg | N/A | |||||
17089343
|
Purolite™
LS01315-192 |
Lifetech™ | 250 g | N/A | |||||
17089353
|
Purolite™
LS01356-94 |
Lifetech™ | 1 kg | N/A | |||||
17099313
|
Purolite™
LS01105-94 |
Lifetech™ | 1 kg | N/A | |||||
17099333
|
Purolite™
LS01245-192 |
Lifetech™ | 250 g | N/A | |||||
17099343
|
Purolite™
LS01315-94 |
Lifetech™ | 1 kg | N/A | |||||
17099363
|
Purolite™
LS01375-192 |
Lifetech™ | 250 g | N/A | |||||
Description
- Enzyme immobilization by adsorption, covalent, IEX and affinity
- High mechanical stability
- High porosity
- Easily dried after immobilization
- Suitable for batch and column configuration
- Optimal for use in organic solvent/water free systems
- Compliant with ResAP(2004) 3
- Halal, Kosher and TSE/BSE/GMO free
Specifications
White to off-white spherical beads | |
Wet | |
Methacrylate (ECR Resin) |
300 to 710 μm | |
Lifetech™ |
Safety and Handling
missing translation for 'shelfLife' : Use within 6 months from date of manufacture