missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Chloride Anhydrous, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
100 g
250 g
5 kg
Specifications
Specifications
Boiling Point | 1382°C |
Melting Point | 605°C |
Color | White |
pH | 7 to 8 |
Physical Form | Solid |
Quantity | 250 g |
Formula Weight | 42.38g/mol |
Packaging | Plastic powder jar |
Vapor Pressure | 1.33 hPa 547 |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling

Product Identifier
- Lithium chloride, anhydrous
- Warning
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/A§.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ A§ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Special Offers
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
Days
28Hours
15Minutes
38Seconds
43
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lithium Chloride Anhydrous, Extra Pure, SLR, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction