missing translation for 'onlineSavingsMsg'
Learn More
Lithium Nitrate Anhydrous, Extra Pure, SLR, Fisher Chemical™ Product Code.: 10568620

Lithium Nitrate Anhydrous, Extra Pure, SLR, Fisher Chemical™

Product Code. 10568620
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 10568620

Brand: Fisher Chemical L/2350/48

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7790-69-4
Molecular Formula LiNO3
Molecular Weight (g/mol) 68.94
MDL Number MFCD00011094
InChI Key IIPYXGDZVMZOAP-UHFFFAOYSA-N
Synonym lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpureShow MoreShow Less
PubChem CID 10129889
ChEBI CHEBI:63315
SMILES [Li+].[O-][N+]([O-])=O

Specifications

Boiling Point 600°C
Melting Point 251°C
Color White
Physical Form Solid
Quantity 100 g
IUPAC Name lithium;nitrate
Formula Weight 68.95g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons
Product Identifier
  • Lithium nitrate
Signal Word
  • Warning
Hazard Category
  • Oxidising solid Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ A§ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lithium Nitrate Anhydrous, Extra Pure, SLR, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.