missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Locator™ Plus Rack and Box Systems

Economically store up to 6,000 samples in the Locator 6 rack and box system that makes sample retrieval simple and consumes minimum liquid nitrogen.

Brand:  Thermo Scientific™ CY50945-70

 View more versions of this product

Product Code. 10398533

  • £5799.00 / Each
Estimated Shipment: 30-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Manual Fill LN2 Dewar
121 L
Vacuum
Without Level Monitor
201 Days
95.3 cm
6 in.
15.2 cm
220 V
2000
Samples (2000)
4.273 cu. ft.
Below -180°C
CE, CSA
Locator 8 Plus
37.5 in.
22 in.
55.9 cm
Alarm, Recorder
0.60L
52.2 kg
8
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Locator™ Plus Rack and Box Systems > Locator 8 Plus; Without Level Monitor

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.