missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Low Serum Growth Supplement (LSGS)

Low Serum Growth Supplement (LSGS)

Brand:  Gibco™ S00310

Product Code. 10616973

  • £89.25 / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Low Serum Growth Supplement (LSGS) is a sterile, concentrated (50X) solution intended for use as one component in a complete culture environment for the growth of human dermal fibroblasts and endothelial cells derived from large vessels. LSGS is intended for use with Medium 106 (fibroblasts) or Medium 200 (endothelial cells). LSGS contains: fetal bovine serum, basic fibroblast growth factor, heparin, hydrocortisone, and epidermal growth factor. When a 500 ml bottle of medium is supplemented with LSGS or the components of the LSGS Kit, the final concentrations of the components in the supplemented medium are:
  • fetal bovine serum, 2% v/v
  • hydrocortisone (1μg/ml)
  • human epidermal growth factor, 10 ng/ml
  • basic fibroblast growth factor, 3 ng/ml
  • heparin, 10μg/ml
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    Store in freezer (-5 to -20°C).
    50 X
    Sterile
    10 mL
    Human
    Epithelial Cells (Large Vessel), Fibroblast Cells (Dermal)
    Growth Supplement
    Cascade Biologics™
    Dry Ice
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates

    Certificates

    A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

    5 results found
    Lot Number Certificate Type Date Catalog Number
    Lot Number2454527 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberS00310
    Lot Number2539428 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberS00310
    Lot Number2790904 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberS00310
    Lot Number3011335 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberS00310
    Lot Number2900617 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberS00310
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Gibco™ Low Serum Growth Supplement (LSGS) > 10mL

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

    For Research Use Only. Not for use in diagnostic procedures.