Learn More
Cytiva HyClone™ HyCell™ CHO Medium, Liquid
Metabolically designed for high cell yield and recombinant protein production
Brand: Cytiva SH30949.02
101.82 GBP valid until 2025-12-31
Use promo code "24275" to get your promotional price.
Description
- ACDF and chemically-defined cell culture media
- High-performance growth in multiple culture systems including high-density bioreactors
- Allows for direct or sequential adaptation
- Supports multiple CHO clones
- Component traceability
- Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
- Sourced from animals that are typically less than 10 days old

Specifications
Manufactured in cGMP (21 CFR 820) Compliant and ISO9001 Certified Facilities | |
With Sodium Bicarbonate/Poloxamer 188, Without HT/L-Glutamine | |
CHO Medium | |
Bottle | |
270 to 330 mOsm/kg | |
HyCell™ |
Clear Yellow | |
Liquid | |
<10 EU/mL | |
Growth Promotion | |
7.0 to 7.4 | |
1 L |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.