Learn More
LRR 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11628-1-AP-150UL
Description
May negatively regulate the 4-1BB-mediated signaling cascades which result in the activation of NK-kappaB and JNK1. Probable substrate recognition subunit of an ECS (Elongin BC- CUL2/5-SOCS-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.Specifications
LRR 1 | |
Polyclonal | |
Unconjugated | |
LRR1 | |
4 1BBLRR, LRR 1, LRR repeat protein 1, LRR1, PPIL5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
122769, 214345, 685860 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q80VQ1, Q96L50 | |
Lrr1, Lrrc1 | |
LRR 1 Fusion Protein Ag2159 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.