Learn More
LRRC18 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24571-1-AP-150UL
 View more versions of this product
Product Code. 16813055
  • £349.00 / 150µL
Explore more special offers
Description
Description
May be involved in the regulation of spermatogenesis and sperm maturation.
Specifications
Specifications
LRRC18
Polyclonal
Unconjugated
LRRC18
LRRC18, UNQ933, UNQ9338, VKGE9338
Rabbit
Antigen Affinity Chromatography
RUO
474354, 67580
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.39 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8N456, Q9CQ07
LRRC18
LRRC18 Fusion Protein Ag20996
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
LRRC18 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated