missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sonics Materials™ Threaded End Probe
Brand: Sonics Materials™ 630-0220
This item is not returnable.
View return policy
Specifications
114 μm | |
13 mm | |
High |
CV188 Converter | |
139 mm | |
1 Ea. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sonics Materials™ Threaded End Probe > Diameter: 13mm
Spot an opportunity for improvement?Share a Content Correction