Learn More
m-Phenylenediamine, 99+%, Thermo Scientific Chemicals
£21.50 - £202.00
Chemical Identifiers
CAS | 108-45-2 |
---|---|
Molecular Formula | C6H8N2 |
Molecular Weight (g/mol) | 108.14 |
MDL Number | MFCD00007799 |
InChI Key | WZCQRUWWHSTZEM-UHFFFAOYSA-N |
Synonym | m-phenylenediamine, 1,3-phenylenediamine, 1,3-diaminobenzene, 1,3-benzenediamine, m-diaminobenzene, 3-aminoaniline, m-aminoaniline, meta-phenylenediamine, developer c, 3-phenylenediamine |
PubChem CID | 7935 |
ChEBI | CHEBI:8092 |
IUPAC Name | benzene-1,3-diamine |
SMILES | C1=CC(=CC(=C1)N)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10563101
|
Thermo Scientific Acros
130560250 |
25 g | Glass bottle |
£21.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10235230
|
Thermo Scientific Acros
130562500 |
250 g | Glass bottle |
£23.40
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10306780
|
Thermo Scientific Acros
130565000 |
500 g | Glass bottle |
£35.70
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10677672
|
Thermo Scientific Acros
130560010 |
1 kg | Glass bottle |
£53.30
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10617753
|
Thermo Scientific Acros
130560050 |
5 kg | Plastic drum |
£202.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
108-45-2 | |
108.14 | |
WZCQRUWWHSTZEM-UHFFFAOYSA-N | |
7935 | |
benzene-1,3-diamine |
C6H8N2 | |
MFCD00007799 | |
m-phenylenediamine, 1,3-phenylenediamine, 1,3-diaminobenzene, 1,3-benzenediamine, m-diaminobenzene, 3-aminoaniline, m-aminoaniline, meta-phenylenediamine, developer c, 3-phenylenediamine | |
CHEBI:8092 | |
C1=CC(=CC(=C1)N)N |
Specifications
108-45-2 | |
100.0 | |
Brown-Gray or Tan to Yellow | |
175°C | |
99% min. (GC) | |
C6H4(NH2)2 | |
03, IV, 79 | |
m-phenylenediamine, 1,3-phenylenediamine, 1,3-diaminobenzene, 1,3-benzenediamine, m-diaminobenzene, 3-aminoaniline, m-aminoaniline, meta-phenylenediamine, developer c, 3-phenylenediamine | |
WZCQRUWWHSTZEM-UHFFFAOYSA-N | |
benzene-1,3-diamine | |
7935 | |
108.14 | |
Pellets |
99.0 | |
63.0°C to 65.0°C | |
282.0°C to 284.0°C | |
Authentic | |
C6H8N2 | |
MFCD00007799 | |
15, 7395 | |
Solubility in water: 350g/L (25°C). Other solubilities: soluble in alcohol,ether and chloroform | |
C1=CC(=CC(=C1)N)N | |
108.14 | |
CHEBI:8092 | |
99+% | |
m-Phenylenediamine |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause an allergic skin reaction.
Causes serious eye irritation.
Suspected of causing genetic defects.
Very toxic to aquatic l
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-584-7
Recommended Storage : this product tends to darken during storage
- m-Phenylenediamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 2
- Respiratory sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H319-Causes serious eye irritation.
- H341-Suspected of causing genetic defects.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only