Learn More
m-Xylylene dichloride, 97%, Thermo Scientific Chemicals
£31.80 - £181.00
Chemical Identifiers
CAS | 626-16-4 |
---|---|
Molecular Formula | C8H8Cl2 |
Molecular Weight (g/mol) | 175.052 |
MDL Number | MFCD00000912 |
InChI Key | GRJWOKACBGZOKT-UHFFFAOYSA-N |
Synonym | 1,3-bis chloromethyl benzene, alpha,alpha'-dichloro-m-xylene, m-xylylene dichloride, benzene, 1,3-bis chloromethyl, m-bis chloromethyl benzene, m-xylylene chloride, m-dichloroxylene, benzene, 1,3-dichloromethyl, ccris 1774, m-xylene, .alpha.,.alpha.'-dichloro |
PubChem CID | 12275 |
IUPAC Name | 1,3-bis(chloromethyl)benzene |
SMILES | C1=CC(=CC(=C1)CCl)CCl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11418790
|
Thermo Scientific Alfa Aesar
A11548.09 |
10 g |
£31.80
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11404583
|
Thermo Scientific Alfa Aesar
A11548.14 |
25 g |
£61.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11414583
|
Thermo Scientific Alfa Aesar
A11548.22 |
100 g |
£181.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
m-Xylylene dichloride is used in pharmaceutical and used in medicine, brightener intermediates. α,α?-Dichloro-m-xylene is suitable for use in the synthesis of 1,7-bis(6-methoxy-2-formylphenyl)-1,7-dioxaheptane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsm-Xylylene dichloride is used in pharmaceutical and used in medicine, brightener intermediates. α,α′-Dichloro-m-xylene is suitable for use in the synthesis of 1,7-bis(6-methoxy-2-formylphenyl)-1,7-dioxaheptane.
Solubility
Insoluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Chemical Identifiers
626-16-4 | |
175.052 | |
GRJWOKACBGZOKT-UHFFFAOYSA-N | |
12275 | |
C1=CC(=CC(=C1)CCl)CCl |
C8H8Cl2 | |
MFCD00000912 | |
1,3-bis chloromethyl benzene, alpha,alpha'-dichloro-m-xylene, m-xylylene dichloride, benzene, 1,3-bis chloromethyl, m-bis chloromethyl benzene, m-xylylene chloride, m-dichloroxylene, benzene, 1,3-dichloromethyl, ccris 1774, m-xylene, .alpha.,.alpha.'-dichloro | |
1,3-bis(chloromethyl)benzene |
Specifications
626-16-4 | |
250°C to 255°C | |
C8H8Cl2 | |
UN2928 | |
1,3-bis chloromethyl benzene, alpha,alpha'-dichloro-m-xylene, m-xylylene dichloride, benzene, 1,3-bis chloromethyl, m-bis chloromethyl benzene, m-xylylene chloride, m-dichloroxylene, benzene, 1,3-dichloromethyl, ccris 1774, m-xylene, .alpha.,.alpha.'-dichloro | |
C1=CC(=CC(=C1)CCl)CCl | |
175.052 | |
175.06 | |
m-Xylylene dichloride |
33°C to 37°C | |
144°C (291°F) | |
MFCD00000912 | |
2079898 | |
GRJWOKACBGZOKT-UHFFFAOYSA-N | |
1,3-bis(chloromethyl)benzene | |
12275 | |
97% |
Safety and Handling
GHS H Statement
H330-H314-H318-H302
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
GHS P Statement
P280-P273-P305+P351+P338-P309-P310-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.
missing translation for 'rtecsNumber' : ZE4070000
missing translation for 'tsca' : TSCA
RUO – Research Use Only