Learn More
Velp Scientifica™ Magnetic Stirring Bars
Emerse this into a liquid to produce a stirring movement. Velp Scientifica™ Magnetic Stirring Bars do not contaminate or react with the mixure they are in.
Brand:  Velp Scientifica™ A00001061
 View more versions of this product
Product Code. 15389634
  • £13.75 / Each
Estimated Shipment: 19-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Emerse this into a liquid to produce a stirring movement. Velp Scientifica™ Magnetic Stirring Bars do not contaminate or react with the mixure they are in.
  • For stirring applications
  • Chemically inert
Specifications
Specifications
VELP Magnetic stirrer, Heating magnetic stirrer
10 mm
60 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Velp Scientifica™ Magnetic Stirring Bars > Length: 60 mm