missing translation for 'onlineSavingsMsg'
Learn More
Maleic anhydride, 98+%, Thermo Scientific Chemicals Product Code.: 11414823

Maleic anhydride, 98+%, Thermo Scientific Chemicals

Product Code. 11414823
1000 g
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy

Product Code. 11414823

Brand: Thermo Scientific Alfa Aesar A12178.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Maleic anhydride is used in the preparation of unsaturated polyester resins, 4-butanediol, fumairc acid, tetrahydrofuran and gamma-butyrolactone. It is also used as lubricating oil additives, dispersants, artificial sweeteners, flavor enhancements, floor polishes and corrosion inhibitors. It finds application in personal care products like hair sprays. Further, it is used in water treatment chemicals, detergents, pharmaceuticals and copolymers. It is an active substrate for Diels-Alder reactions and dimerizes to form cyclobutane tetracarboxylic dianhydride .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Maleic anhydride is used in the preparation of unsaturated polyester resins, 4-butanediol, fumairc acid, tetrahydrofuran and gamma-butyrolactone. It is also used as lubricating oil additives, dispersants, artificial sweeteners, flavor enhancements, floor polishes and corrosion inhibitors. It finds application in personal care products like hair sprays. Further, it is used in water treatment chemicals, detergents, pharmaceuticals and copolymers. It is an active substrate for Diels-Alder reactions and dimerizes to form cyclobutane tetracarboxylic dianhydride .

Solubility
Soluble in water, acetone and dioxane. Slightly soluble in methanol, chloroform and benzene.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases, strong reducing agents, alkali metals and amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-31-6
Molecular Formula C4H2O3
Molecular Weight (g/mol) 98.06
MDL Number MFCD00005518
InChI Key FPYJFEHAWHCUMM-UHFFFAOYSA-N
Synonym maleic anhydride, 2,5-furandione, maleic acid anhydride, toxilic anhydride, dihydro-2,5-dioxofuran, cis-butenedioic anhydride, 2,5-dihydrofuran-2,5-dione, polymaleic anhydride, maleinanhydrid, rcra waste number u147Show MoreShow Less
PubChem CID 7923
ChEBI CHEBI:474859
IUPAC Name furan-2,5-dione
SMILES O=C1OC(=O)C=C1

Specifications

Melting Point 52°C to 56°C
Boiling Point 202°C
Flash Point 103°C (217°F)
Assay Percent Range ≥98%
Quantity 1000 g
UN Number UN2215
Beilstein 106909
Sensitivity Moisture sensitive
Merck Index 14,5704
Solubility Information Soluble in water,acetone and dioxane. Slightly soluble in methanol,chloroform and benzene.
Formula Weight 98.06
Percent Purity ≥98%
Chemical Name or Material Maleic Anhydride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253183 Certificate TypeCertificate of Analysis Date25/12/2024 Catalog NumberA12178.0B,A12178.0I,A12178.30
Lot Number10250754 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA12178.0B,A12178.0I,A12178.30
Lot Number10249141 Certificate TypeCertificate of Analysis Date01/01/2024 Catalog NumberA12178.0B,A12178.0I,A12178.30
Lot Number10237465 Certificate TypeCertificate of Analysis Date07/11/2023 Catalog NumberA12178.0B,A12178.0I,A12178.30
Lot Number10201196 Certificate TypeCertificate of Analysis Date07/11/2023 Catalog NumberA12178.0B,A12178.0I,A12178.30
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Maleic anhydride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H372-Causes damage to organs.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Maleic anhydride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Maleic anhydride, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.