missing translation for 'onlineSavingsMsg'
Learn More
Manganese(II) chloride tetrahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11306917

Manganese(II) chloride tetrahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11306917
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11306917

Brand: Thermo Scientific Alfa Aesar 010804.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Manganese chloride is used as a source of manganese ion in biological research, such as in studies of manganese uptake in rat liver. It has been substituted for magnesium chloride to investigate alterations in activity of the Herpes simplex virus type 1 DNA polymerase. It is also used in industrial applications such as dyeing and disinfecting, the purification of natural gas, and as a drying agent for linseed oil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-34-9
Molecular Formula Cl2H8MnO4
Molecular Weight (g/mol) 197.90
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
IUPAC Name manganese(2+) tetrahydrate dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]

Specifications

Melting Point 58°C
Color Pink
Physical Form Crystalline Powder
Quantity 10 g
Assay Percent Range 99.999% (Metals basis)
Solubility Information Soluble in water, pyridine and ethanol. Insoluble in ether.
Formula Weight 197.90 (125.84 Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Density 2.010 g/cm3 at 20°C
Chemical Name or Material Manganese(II) chloride tetrahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP16H094 Certificate TypeCertificate of Analysis Date09/01/2025 Catalog Number010804.18,010804.09
Lot NumberS07I011 Certificate TypeCertificate of Analysis Date20/06/2022 Catalog Number010804.18,010804.09
Lot Number24049 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number010804.18,010804.09
Lot Number23343 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number010804.18,010804.09
Lot Number23352 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number010804.18,010804.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Manganese(II) chloride tetrahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.