Learn More
Manganese(II) oxide, Mn 76.0-78.0%, typically 99%, Thermo Scientific Chemicals
Description
The main use of manganese dioxide is in dry-cell batteries, such as the alkaline battery and the zinc-carbon battery. It is useful as a pigment, and as a precursor to other manganese compounds. It is used as a reagent in organic synthesis, for instance, for the oxidation of allylic alcohols and arylmethyl alcohols. It is also employed in various industrial purposes such as in adhesives and sealant chemicals, lubricants and lubricant additives, fertilizers, oxidizing/reducing agents, plating agents and surface treating agents, processing aids, air care products, paints, and coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Green |
Physical Form | Powder |
Quantity | 2 kg |
Assay Percent Range | Mn 76.0-78.0%, typically 99% |
Solubility Information | Insoluble in water. Soluble in hydrochloric acid. |
Formula Weight | 70.94 |
Odor | Odorless |
Appearance | Green powder |
Density | 5 |
Chemical Name or Material | Manganese(II) oxide, Mn 76.0-78.0% |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Z05J021 | Certificate of Analysis | 04/01/2024 | 011870.F7,011870.A3,011870.36 |
R24J024 | Certificate of Analysis | 06/06/2023 | 011870.A3 |
Z29I019 | Certificate of Analysis | 09/01/2023 | 011870.F7,011870.A3,011870.36 |
Z02H045 | Certificate of Analysis | 12/04/2022 | 011870.F7,011870.A3,011870.36 |
A17Q38 | Certificate of Analysis | 07/12/2021 | 011870.F7,011870.A3,011870.36 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.