missing translation for 'onlineSavingsMsg'
Learn More
Manganese(II) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11337567

Manganese(II) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11337567
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11337567

Brand: Thermo Scientific Alfa Aesar 012835.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

it is used as an intermediate in chemical research& pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18820-29-6
Molecular Formula MnS
Molecular Weight (g/mol) 87.00
MDL Number MFCD00014210
InChI Key VCTOKJRTAUILIH-UHFFFAOYSA-N
Synonym manganese sulfide, manganese ii sulfide, manganous sulfide, manganese sulphide, manganese 2+ sulfide, 12687-82-0 cpd with unspecified mf, manganese ii sulfide, ∼100 mesh particle sizeShow MoreShow Less
PubChem CID 87809
SMILES [S--].[Mn++]

Specifications

Melting Point 1610°C (decomposition)
Color Green
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.9% (Metal basis)
Solubility Information It is soluble in water. 0.47mg/100 mL (18°C).
IUPAC Name manganese(2+) sulfanediide
Formula Weight 87
Appearance Green powder
Density 3.99
Chemical Name or Material Manganese(II) sulfide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP25L004 Certificate TypeCertificate of Analysis Date27/03/2025 Catalog Number012835.14,012835.06,012835.22
Lot NumberN05K031 Certificate TypeCertificate of Analysis Date16/02/2024 Catalog Number012835.14,012835.06,012835.22
Lot NumberW25J041 Certificate TypeCertificate of Analysis Date04/10/2023 Catalog Number012835.14,012835.06,012835.22
Lot NumberR23I031 Certificate TypeCertificate of Analysis Date01/06/2022 Catalog Number012835.14,012835.06,012835.22
Lot NumberN10I012 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number012835.14,012835.06,012835.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Manganese(II) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.