missing translation for 'onlineSavingsMsg'
Learn More

Manganese(IV) oxide, 99%, -10 mesh, Thermo Scientific Chemicals

£19.40 - £279.39

Chemical Identifiers

CAS 1313-13-9
Molecular Formula MnO2
Molecular Weight (g/mol) 86.94
MDL Number MFCD00003463
InChI Key NUJOXMJBOLGQSY-UHFFFAOYSA-N
Synonym manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
PubChem CID 14801
IUPAC Name dioxomanganese
SMILES O=[Mn]=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11984141
View Documents
Thermo Scientific Acros
222581000
100 g Plastic bottle
£19.40
100g
Please to purchase this item. Need a web account? Register with us today!
11994141
View Documents
Thermo Scientific Acros
222585000
500 g Plastic bottle
£62.61
500g
Please to purchase this item. Need a web account? Register with us today!
11974141
View Documents
Thermo Scientific Acros
222580025
2.5 kg Plastic Bottle
£279.39
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Chemical Identifiers

1313-13-9
86.94
NUJOXMJBOLGQSY-UHFFFAOYSA-N
14801
O=[Mn]=O
MnO2
MFCD00003463
manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
dioxomanganese

Specifications

535.0°C
99%
MnO2
1479
15,5795
Solubility in water: insoluble. Other solubilities: soluble in hydrochloric acid
O=[Mn]=O
86.94
86.94
5.02
1313-13-9
MnO2
MFCD00003463
01,637; 02,257; 03,191; 04,317; 05,422; 06,357; 08,312; 13,233; 14,200; 15,197; 16,202
manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
NUJOXMJBOLGQSY-UHFFFAOYSA-N
dioxomanganese
14801
99%
Manganese(IV) oxide, 99+%, -100 mesh
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure if inhaled.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
D

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 215-202-6

missing translation for 'rtecsNumber' : OP0350000

missing translation for 'tsca' : TSCA

Product Identifier
  • Manganese(IV) oxide

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contains : Manganese oxide

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only