missing translation for 'onlineSavingsMsg'
Learn More
Manganese(IV) oxide, Puratronic™, 99.996% (metals basis), Thermo Scientific Chemicals Product Code.: 11326917

Manganese(IV) oxide, Puratronic™, 99.996% (metals basis), Thermo Scientific Chemicals

Product Code. 11326917
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11326917

Brand: Thermo Scientific Alfa Aesar 010805.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used as an oxidizing agent in organic synthesis such as oxidation of allylic/benzylic alcohols, as a textile dye, as a reducing agent, and as a component of dry cell batteries such as zinc-carbon battery and alkaline battery. It is also used in making pigments for glasses and ceramics, and drier for paints. Further it is used in the manufacture of manganese steel and several manganese derivatives including potassium permanganate, a powerful versatile oxidant. Catalytic hydration of nitriles to amides by flowing through manganese dioxide has been reported. Manganese dioxide supported on inorganic oxide can be used for oxidation of methylamine through CWAO (Catalytic Wet Air Oxidation). It has high potential as highly efficient and robust material for water oxidation reaction (WORs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-13-9
Molecular Formula MnO2
Molecular Weight (g/mol) 86.94
MDL Number MFCD00003463
InChI Key NUJOXMJBOLGQSY-UHFFFAOYSA-N
Synonym manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunsteinShow MoreShow Less
PubChem CID 14801
IUPAC Name dioxomanganese
SMILES O=[Mn]=O

Specifications

Melting Point 535°C (decomposition)
Color Brown
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.997% (Metals basis)
Solubility Information Insoluble in water
Formula Weight 86.94
Odor Odorless
Density 5.026
Chemical Name or Material Manganese(IV) oxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ02K010 Certificate TypeCertificate of Analysis Date04/12/2024 Catalog Number010805.06,010805.14,010805.22
Lot NumberU07K061 Certificate TypeCertificate of Analysis Date16/08/2024 Catalog Number010805.06,010805.14,010805.22
Lot NumberQ26K046 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number010805.06,010805.14,010805.22
Lot Number10247414 Certificate TypeCertificate of Analysis Date19/07/2023 Catalog Number010805.06,010805.14,010805.22,010805
Lot NumberX31I046 Certificate TypeCertificate of Analysis Date11/11/2022 Catalog Number010805.06,010805.14,010805.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Manganese(IV) oxide, Puratronic™, 99.996% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.