missing translation for 'onlineSavingsMsg'
Learn More
Manganese sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis), Thermo Scientific Chemicals Product Code.: 15414805

Manganese sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 15414805
1 Ea.
Click to view available options
Quantity:
1 Ea.
This item is not returnable. View return policy

Product Code. 15414805

Brand: Thermo Scientific Alfa Aesar 040864.KS

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

Manganese sputtering target is widely used in coating processing industries like magnetic data storage application, electronic and semiconductor application. It is used also in chemical vapor deposition (CVD), physical vapor deposition (PVD) display and optical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Ea.
CAS 7439-96-5
Form Hot Pressed
Odor Odorless
Molecular Formula Mn
MDL Number MFCD00011111
Synonym colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds
Solubility Information Insoluble in water.
InChI Key PWHULOQIROXLJO-UHFFFAOYSA-N
SMILES [Mn]
IUPAC Name manganese
Molecular Weight (g/mol) 54.94
PubChem CID 23930
ChEBI CHEBI:35154
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Manganese sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA10Y010 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040864.KS
Lot NumberC30Q05 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040864.KS
Lot NumberI06Q44 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040864.KS
Lot NumberI08P13 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number040864.KS
Lot NumberR04B050 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number040864.KS

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Manganese sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.