Learn More
Masterflex™ I/P™ Brushless Process Drive with High-Performance Pump Head for High-Performance Precision Tubing
Powerful, low-maintenance drives are versatile and cost-effective
Brand:  Masterflex™ 77963-20
Product Code. 11876883
Description
Description
  • Digital interface with LED display shows percent speed with 1% resolution
  • Speed control via three-turn potentiometer for quick, easy adjustments
  • Easily control start/stop and reverse features via sealed keypad
  • Constructed of non-chip painted steel and aluminum IP55-rated housing
Specifications
Specifications
CE, cULus, ISO 9001:2008
LED
17 L/min.
Epoxy-Coated
06429-88, 3 ft. (1m) Tubing, 77410-10 Drive, I/P High-Performance 77600-62 Pump Head, Tygon LFLI/P88
I/P Precision Brushless
I/P High-Performance pump head 77600-62
355.6 mm
3.5/1.8 A 115/230 VAC 50/60 Hz
50/60 Hz
Variable Speed
IP55
650 rpm
1/3 hp
Aluminum, Steel
1/3 hp brushless
416.05 mm
228.6 mm
90/260 VAC
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Masterflex™ I/P™ Brushless Process Drive with High-Performance Pump Head for High-Performance Precision Tubing > Flow Rate: 0.7 to 17LPM