missing translation for 'onlineSavingsMsg'
Learn More

Cytiva MBPTrap™ HP Prepacked Columns

Yield highly pure MBP-tagged recombinant proteins eluted in concentrated form and small volumes. Cytiva Life Sciences™ MBPTrap™ HP Prepacked Columns are prepacked with Dextrin Sepharose™ High Performance.

Brand:  Cytiva 28-9187-80

Code : GE

Additional Details : Weight : 0.12870kg


 View more versions of this product

Product Code. 11500664

  • £935.00 / Pack of 5
Estimated Shipment: 02-09-2024
to see stock.
Add to basket

Description

Description

  • Prepacked with Dextrin Sepharose tagged with maltose binding protein (MBP).
  • Binding capacity is fusion protein dependent but can be as much as 16 mg/ml media
  • Maximum pressure: 3 bar
  • Compatible with commonly used buffers and easily regenerated with 0.5 M NaOH
  • 1 and 5mL polypropylene columns can be used either manually using a syringe or on an instrument

Media
  • Matrix: 6% cross-linked agarose
  • Average particle size: 34μm
  • pH stability working range: >7

1mL columns

  • Bed dimensions: 7 x 25mm
  • Flow velocity: <4mL/min.

5mL columns
  • Bed dimensions: 16 x 25mm
  • Flow velocity: <20mL/min.

Specifications

Specifications

Aqueous Buffer
1mL
16mm
0.98 in.
<20mL/min.
Dextrin
>7
6% Highly Cross-linked Agarose
Ion Exchange Column
Dextrin Sepharose HP prepacked column
0.62 in.
5 x 5mL
25mm
Polypropylene
3 bar
34μm
2° to 8°C
Purification of MBP tagged Proteins
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • MBPTrap HP

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers