missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva MBPTrap™ HP Prepacked Columns
Yield highly pure MBP-tagged recombinant proteins eluted in concentrated form and small volumes. Cytiva Life Sciences™ MBPTrap™ HP Prepacked Columns are prepacked with Dextrin Sepharose™ High Performance.
£229.00 - £935.00
Specifications
Buffer | Aqueous Buffer |
---|---|
Product Type | Dextrin Sepharose HP prepacked column |
Bed Volume | 1mL |
Height (English) | 0.98 in. |
Height (Metric) | 25mm |
Product Code | Brand | Flow Rate | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Flow Rate | Quantity | Price | Quantity & Availability | ||||
10097924
|
Cytiva
28-9187-79 |
<20mL/min. | 1 x 5mL |
£229.00
Each |
Estimated Shipment: 18-09-2024
Log in to see stock. |
||||
10075174
|
Cytiva
28-9187-78 |
<4mL/min. | 1 x 5mL |
£252.00
Pack of 5 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
11500664
|
Cytiva
28-9187-80 |
<20mL/min. | 5 x 5mL |
£935.00
Pack of 5 |
Estimated Shipment: 30-08-2024
Log in to see stock. |
||||
Description
- Prepacked with Dextrin Sepharose tagged with maltose binding protein (MBP).
- Binding capacity is fusion protein dependent but can be as much as 16 mg/ml media
- Maximum pressure: 3 bar
- Compatible with commonly used buffers and easily regenerated with 0.5 M NaOH
- 1 and 5mL polypropylene columns can be used either manually using a syringe or on an instrument
Media
- Matrix: 6% cross-linked agarose
- Average particle size: 34μm
- pH stability working range: >7
1mL columns
- Bed dimensions: 7 x 25mm
- Flow velocity: <4mL/min.
5mL columns
- Bed dimensions: 16 x 25mm
- Flow velocity: <20mL/min.
Specifications
Aqueous Buffer | |
1mL | |
25mm | |
Dextrin | |
>7 | |
6% Highly Cross-linked Agarose | |
Ion Exchange Column |
Dextrin Sepharose HP prepacked column | |
0.98 in. | |
Polypropylene | |
3 bar | |
34μm | |
2° to 8°C | |
Purification of MBP tagged Proteins |