Learn More
Lovibond™ MD 100 Chlorine Tablet
Lovibond MD 100 Chlorine Tablet, Applications: Testing in the field, laboratory or plant room, Detection Range: 0-6 - 0-10mg/L, Display Type: LCD, Filling Solution Type: Reagent
Brand:  Lovibond™ 276000
Product Code. 12641705
  • £520.00 / Each
Explore more special offers
Description
Description
Lovibond MD 100 Chlorine Tablet, Applications: Testing in the field, laboratory or plant room, Detection Range: 0-6 - 0-10mg/L, Display Type: LCD, Filling Solution Type: Reagent
Specifications
Specifications
MD 100 Chlorine Tablet
LCD
5°C to 40°C
4 Micro batteries (AAA), 3 round vials (Glass) with lids, tablet reagents and/or liquid reagents, guarantee sheet, certificate (COC), instruction manual
155 mm
0 to 6 - 0 to 10 mg/L
Reagent
Testing in the field, laboratory or plant room
5 or 10 Year
75 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Brochures
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lovibond™ MD 100 Chlorine Tablet > Applications: Testing in the field, laboratory or plant room