missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Medium 199 (2X)
GREENER_CHOICE

Medium 199 was originally developed for nutritional studies of chick embryo fibroblasts. It has broad species applicability, particularly for cultivation of non-transformed cells.

Brand:  Gibco™ 21157029

Product Code. 11590556

  • £59.00 / 250mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Medium 199 was originally developed for nutritional studies of chick embryo fibroblasts. It has broad species applicability, particularly for cultivation of non-transformed cells. Medium 199 is widely used in virology, vaccine production, and in vitro cultivation of primary explants of mouse pancreatic epithelium, and rat lens tissues. We offer a variety of Gibco™ Medium 199 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This Medium 199 is modified as follows:

With: L-glutamine

Without: HEPES, Phenol Red

The complete formulation is available.

Compared to other basal media, Medium 199 contains unique components, including adenine, adenosine, hypoxanthine, thymine, and additional vitamins. Medium 199 is available with Earle's salts for use in a CO2 incubator, or with Hanks' salts for use without CO2. This Medium 199 is made with Earle's salts.

Medium 199 contains no proteins, lipids, or growth factors. Medium 199 uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: 2 – 8°C. Protect from light.
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
2 X
Liquid
Sterile-filtered
No HEPES, No Phenol Red
Gibco™
Room Temperature
Mammalian Cells
Mammalian Cell Culture
Medium 199
Glutamine, Earle's Balanced Salt Solution (EBSS)
cGMP-compliant under the ISO 13485 standard
250 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3062678 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number21157029
Lot Number2715522 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number21157029
Lot Number2829085 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog Number21157029
Lot Number2917316 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number21157029
Lot NumberA0463723 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number211570010,211570051,21157029,211572500
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Medium 199 (2X) > Quantity: 250 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.