missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Medium Two Prong Sleeves

OHAUS™ Medium Two Prong Sleeves are for use with clamps and can be easily removed for cleaning or replacement.

Brand:  OHAUS™ 30400253

8.89 GBP valid until 2025-06-13
Use promo code "24123" to get your promotional price.


 View more versions of this product

Product Code. 15722669

  • £10.35 / Each
Estimated Shipment: 27-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Available in red vinyl and fiberglass
  • Fiberglass sleeves are ideal for application above 100°C
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2
Vinyl
Red
Prong Sleeve
Medium
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
OHAUS™ Medium Two Prong Sleeves > Material: Vinyl

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.