Learn More
Fisherbrand™ Meker Burner, Natural Gas
Fitted with a perforated steel head, which gives off multiple small but very hot flames.
Brand: Fisherbrand™ 4101/32
Description
- The hottest flame temperature for all natural gas burners is between 900° and 1000°C at the tip of the blue flame cone
- Constructed utilizing a heavy pressure diecast base painted in a blue chemically resistant acrylic finish
- All burners tested for flame quality and leakage prior to packing
- Tests are carried out at the standard gas pressure for each specific type gas
- Temperatures dependent on mains pressure and the chemical construction of the gas which varies in different parts of the world
- Standard mains pressure for natural gas is 20 millibars
- Not fitted with an air regulator
Specifications
Meker Burner | |
30 mm | |
1 Ea. |
Natural Gas | |
192 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Product Content Correction
Product Title
Fisherbrand™ Meker Burner, Natural Gas > Gas Type: Natural/Coal
Spot an opportunity for improvement?Share a Content Correction