Learn More
Melamine, 99%, Thermo Scientific Chemicals
Description
Melamine is used in the production of synthetic resins such as melamine resin with formaldehyde, which is used in high pressure decorative laminates like formica, melamine dinnerware, laminate flooring, and dry erase boards. The presence rich in nitrogen content resulted in fire retardant properties when mixed with resins. It is used to prepare melamine cyanurate by reacting with cyanuric acid. It finds application as superplasticizer for making high-resistance concrete, soundproofing material and in polymeric cleaning products like magic eraser. It is an active component in pigment yellow 150. Further, it is also used as a nitrogen and carbon source for N-doped carbon nanotube.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Melamine is used in the production of synthetic resins such as melamine resin with formaldehyde, which is used in high pressure decorative laminates like formica, melamine dinnerware, laminate flooring, and dry erase boards. The presence rich in nitrogen content resulted in fire retardant properties when mixed with resins. It is used to prepare melamine cyanurate by reacting with cyanuric acid. It finds application as superplasticizer for making high-resistance concrete, soundproofing material and in polymeric cleaning products like magic eraser. It is an active component in pigment yellow 150. Further, it is also used as a nitrogen and carbon source for N-doped carbon nanotube.
Solubility
Slightly soluble in water, hot alcohol, glycerol, glycol and pyridine. Insoluble in ether, benzene and carbon tetrachloride.
Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong acids.

Specifications
Specifications
Melting Point | ∼350°C (decomposition) |
Density | 1.573 |
Quantity | 2500 g |
Beilstein | 124341 |
Merck Index | 14,5811 |
Solubility Information | Slightly soluble in water,hot alcohol,glycerol,glycol and pyridine. Insoluble in ether,benzene and carbon tetrachloride. |
Formula Weight | 126.12 |
Percent Purity | 99% |
Chemical Name or Material | Melamine |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N18L042 | Certificate of Analysis | 20/02/2025 | A11295.36,A11295.0C,A11295.0E |
Y27K026 | Certificate of Analysis | 02/12/2024 | A11295.0C,A11295.36,A11295.0E |
10251735 | Certificate of Analysis | 30/08/2024 | A11295.0C,A11295.36,A11295.0E |
10251840 | Certificate of Analysis | 30/08/2024 | A11295.0C,A11295.36,A11295.0E |
10251305 | Certificate of Analysis | 07/08/2024 | A11295.0C,A11295.36,A11295.0E |
Safety and Handling

- Warning
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
- H351-Suspected of causing cancer.
- H361f-Suspected of damaging fertility.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P235-Keep cool.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.